ActivArmr® 28-360 Industrial Gloves, Ansell
Supplier: Ansell Healthcare
216151 216152 216153
80092-702CS 3629.84 CAD
80092-702 80092-704 80092-706
ActivArmr® 28-360 Industrial Gloves, Ansell
Gloves
In light oil applications, the fully coated 28-360 gloves provide better grip and cut protection (ANSI Cut Level 2) than ordinary cut-resistant knit gloves.
  • Porous coating channels oil away effectively
  • Superior flexibility
  • Great resistance to cuts, punctures, and abrasions
The patented porous nitrile coating channels oil away from the surface for better grip, while providing superior flexibility and dexterity, and resists cuts, punctures, and abrasions.
Use in the appliance manufacturing, automotive, general industry, machining, and metal fabrication industries. Uses include handling abrasive or sharp-edged building materials; handling door panels, hoods and other sharp-edged materials; handling glass or slippery plastics; heavy equipment assembly and cleanup; heavy-duty handling of castings, cores and fabricated metals; servicing and maintaining metalworking equipment; and sheet metal handling and finishing.
The five-piece jersey lining incorporates cut-resistant Kevlar® panels in the high-stress areas.
Certifications: ANSI level 2 cut protection. ANSI level 3 abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 6 bags in a carton.