ActivArmr® 28-359 Cut resistant gloves, Ansell
Supplier: Ansell Canada
2835911080 2835911090
CA32890-464CS 1788.45 CAD
CA32890-464 CA32890-466 CA32890-468
ActivArmr® 28-359 Cut resistant gloves, Ansell
Gloves
The standard of safety and grip when working in oily environments, the fully coated 28-359 gloves resist cuts nine times better than leather-palm designs, with twice the puncture resistance and 20 times the abrasion resistance.
  • Ergonomically designed for comfortable use
  • Increased flexibility
  • Exceptional resistance to cuts
The 28-359 offers superior dry grip and the nitrile coating repels oil and grease.
Use in the aerospace, appliance manufacturing, automotive, general industry, HVAC/Sheet Metal Users/Window Glazers, metal fabrication, plumbers, public utilities, appliance manufacturing, and automotive industries. Uses include handling abrasive or sharp-edged building materials; handling and assembling; HVAC components; handling door panels, hoods and other sharp-edged materials; handling heavy metals and trimming/cutting plastic, carpet and cardboard; heavy-duty handling of castings, cores and fabricated metals; and sheet metal handling and finishing.
Gloves are ergonomically designed with a five-piece jersey lining for increased flexibility and worker comfort.
Certifications: Complies with FDA regulation 21 CFR 177.2600 for food contact. ANSI level 3 cut protection; ANSI Level 4 Abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 6 bags in a carton.