AlphaTec® 09-924 Chemical resistant gloves, Ansell
Supplier: Ansell
AlphaTec®
213290
80079-956CS 3782.52 CAD
80079-956
AlphaTec® 09-924 Chemical resistant gloves, Ansell
Gloves
Gloves combine the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner.
  • Excellent thermal and chemical protection
  • Great abrasion resistance
  • Protect against greases and oils
  • Seamless design eliminates discomfort
  • Superior flexibility and grip
They are designed to protect user in applications where thermal and chemical risks are present.
Uses include chemical plants and transportation; circuit board manufacturing; handling equipment and abrasive materials coated with oil, grease, acids and solvents; handling oils, acids, caustics and alcohols; chemical processing, oil refining and petrochemicals; detergents and cleaning liquids; handling of objects in hot or cold chemicals; and pesticide manufacturing.
Unaffected by detergents and cleaning solutions. Comfortable 2-piece lining eliminates irritating seams and is suitable for intermittent handling of warm liquids. Provides protection from intermittent contact with hot surfaces up to 180 °C.
Certifications: Compliant with FDA food handling regulation 21 CRF 177.2600. ANSI level 2 cut protection. ANSI level 3 abrasion protection.
Packaging: Gloves are packaged 12 pair per bag; 6 bags in carton.
Caution: Do NOT use the gloves in or around open flame.