HyFlex® 74-301 Three-Strand Stainless Steel Gloves, White
Supplier: Ansell
HyFlex®
222061 222063 222062 222064 222159
80092-792CS 633.09 CAD
80092-792 80092-728 80092-726 80092-725 80092-722
HyFlex® 74-301 Three-Strand Stainless Steel Gloves, White
Gloves
The 74-301 gloves offer an exceptionally high protection from cuts with the power of steel at a more cost-effective price than chain mesh designs.
  • Exceptional cut resistance
  • Reversible design for use on either hand
  • Seamless for added comfort
  • Machine washable for reuse
The cut resistance technology of the 74-301 features a blend of fibers and advanced knitting methods that provide the protection of stainless steel with a seam-free design to offer added comfort.
Use in the food processing, food service, automotive, machining, masonry and tile, metal fabrication, metalworking, paper, plastics, and textiles industries. Uses include beef, pork, poultry, and fish processing; food service; kill and processing areas; fruit and vegetable processing (sorting, cleaning, paring); and as a liner for freehand glove in de-boning areas.
Certifications: ANSI level 8 cut protection. ANSI level 3 abrasion protection.
Packaging: Gloves are packaged 12 each per case.