HyFlex® 74-048 High Cut Protection Gloves, Ansell
Supplier: Ansell Healthcare
245311 245312
80093-042CS 750.38 CAD
80093-042 80093-044
HyFlex® 74-048 High Cut Protection Gloves, Ansell
Gloves
The 74-048 white Dyneema® gloves provide the superior protection of a heavyweight glove and a close, comfortable fit like a lightweight glove.
  • Excellent levels of Cut protection ANSI Level 6
  • Incorporates Tuff-Cuff™ anti-shrink cuff construction
  • Excellent Protection in Food processing applications
  • Good for most knife-hand and some offhand applications
For most knifehand and some offhand applications, gloves provide protection that bears up under even the most adverse conditions.
Gloves deliver unparalleled protection and performance within the Food Processing industry in applications such as beef, pork, poultry, seafood, vegetable, and fruit processing. Also used in the food service, beverage manufacturing, fishing, and agriculture industries. Additional uses include canning; cleaning and changing stationary slicer blades in delis and supermarkets; and all types of processing areas.
In addition, an extended cuff offers extra cut protection around the sensitive wrist area. Manufactured from Dyneema® these gloves feature Tuff-Cuff™ technology, a revolutionary development that dramatically reduces cuff blowout even after multiple launderings in bleach.
Certifications: The gloves feature ASTM-ISEA Level 5 cut resistance, the highest level of cut resistance in the ASTM-ISEA standard.
Packaging: Gloves are packaged 12 each per case.