AlphaTec® 62-400 Fully Coated Natural Rubber Latex Gloves with Cotton Liner, Ansell
Supplier: Ansell Healthcare
285652 285653 285660
80093-106CS 1590.92 CAD
80093-106 80093-108 80093-110 80093-112
AlphaTec® 62-400 Fully Coated Natural Rubber Latex Gloves with Cotton Liner, Ansell
Gloves
Flexible and durable chemical-protection gloves that are useful for a wide range of jobs.
  • Great flexibility and durability
  • Cotton liner keeps hands comfortable in hot or cold environments
  • Excellent grip for wet applications
  • Gloves stay flexible in cold temperatures
Fully coated natural rubber latex gloves offer great flexibility and durability.
Use in the chemical, fishing, food processing and service, general assembly, janitorial services, and petrochemical industries. Uses include chemical plants and transportation; chemical processing, oilMore Product Information