LC Autosampler Syringes, CTC, SGE
Supplier: Trajan Scientific and Medical
005331 004810 005330 006330 002710 003715 005715 005720 007720
14225-316EA 194.81 CAD
14225-316 14233-484 60362-198 60362-204 89238-036 89238-040 89238-046 89238-048 89238-052
LC Autosampler Syringes, CTC, SGE
Syringes Syringes for Chromatography
SGE has been making precision chromatography syringes for over 50 years
Features and Benefits–Unique plunger design minimizes bending and seizing.
Metal plungers are individually fitted to the syringe glass barrel for a perfect “feel”, optimized life with minimal carry over, a liquid tight seal between the barrel and plunger and excellent performance.
PTFE tipped replaceable plunger syringes reduce the risk of plunger seizing when working with dirty samples, reducing instrument downtime, and minimizing the risk of instrument shutdown during the processing of large batches of samples.
Accuracy and Reproducibility is ± 1 % (displaced volume). Temperature Range is 5-70 °C Fixed Needle, 5-120 °C Removable Needle.
Our range of HPLC autosampler syringes are designed and tested to meet critical autosampler specifications.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Range of fixed needle and removable needle options available. PTFE tipped replaceable plunger versions also available.