L(+)-Asparagine, anhydrous 98,0-102,0, bioreagent excipient_suitable

Supplier: Avantor Performance Materials

Synonyms: L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH

VWRB15005-1KG VWRB15005-25KG VWRB15005-5KG
VWRB15005-1KGEA 704.13 CAD
VWRB15005-1KG VWRB15005-25KG VWRB15005-5KG
L(+)-Asparagine, anhydrous 98,0-102,0, bioreagent excipient_suitable
L(+)-Asparagine

Suitable for bioprocessing. BSE/TSE Free. Manufactured in USA. Custom Packaging and Testing Available. Change Management. Sample availability.

Formula: H₂NCOCH₂CH(NH₂)CO₂H
MW: 132.12 g/mol
Boiling Pt: 343
Melting Pt: 234…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00064401
CAS Number: 70-47-3
EINECS: 200-735-9
Merck Index: 12,00872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance white crystalline powder
Assay (on dried basis, by titration) 98.0% to 102.0%
Identification (by IR) conforms to structure
Aspartic Acid Content NMT 2%
Loss On Drying (@105ºC/3hrs.) NMT 0.5%
Residue On Ignition NMT 0.1%
Solubility (10% solution in dilute HCl) clear, complete, and colorless
Specific Rotation (on dried basis, C=10, 6N HCl) +33.0º to +36.5º
Bioburden NMT 100 CFU/g
Heavy Metals (as Pb) NMT 15 ppm
Chloride (Cl) NMT 0.02%
Sulfate (SO4) NMT 0.03%
Cytotoxicity passes test
Endotoxin (measured at 2g/L) . NMT 0.04 EU/mg
DNase none detected
RNase none detected
Protease none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR