4-Nitrophenol ≥99.5% indicator, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
1-Hydroxy-4-nitrobenzene, p-Nitrophenol
1.06798.0100
1.06798.0025
1.06798.0100
EM1.06798.0025CS
26362.58
CAD
EM1.06798.0025
EM1.06798.0100
CA1.06798.0025
CA1.06798.0100
4-Nitrophenol ≥99.5% indicator, Supelco®
4-Nitrophenol
4-Nitrophenol indicator.
Formula:
C₆H₅NO₃ Boiling Pt: 279 °C (1013 hPa) Melting Pt: 110…114 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: 169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007331 CAS Number: 100-02-7 EINECS: 202-811-7 UN: 1663 ADR: 6.1,III Merck Index: 13,06654 |
Specification Test Results
Hill Formula | C₆H₅NO₃ |
Chemical formula | O₂NC₆H₄OH |
Molar Mass | 139.11 g/mol |
Boiling point | 279 °C (1013 hPa) (decomposition) |
Density | 1.48 g/cm³ (20 °C) |
Flash point | 169 °C |
Ignition temperature | 510 °C |
Melting Point | 110 - 114 °C |
pH value | 4.4 (5 g/l, H₂O, 24 °C) |
Bulk density | 550 kg/m³ |
Solubility | 16 g/l (25 °C) |
LD 50 dermal | LD50 Rat 1024 mg/kg |
Storage class | 6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects |
WGK | WGK 2 water endangering |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...