Sodium-L(+)-glutamate monohydrate, EMPROVE® ESSENTIAL FCC, E621, NF, SAFC®

Supplier: MilliporeSigma

Synonyms: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate

1.06445.1000 1.06445.9029 1.06445.9029
CA1.06445.1000EA 248.21 CAD
CA1.06445.1000 CA1.06445.9029 EM1.06445.9029
Sodium-L(+)-glutamate monohydrate, EMPROVE® ESSENTIAL FCC, E621, NF, SAFC®
Sodium-L(+)-glutamate monohydrate

Sodium L-glutamate monohydrate suitable for use as excipient EMPROVE® exp FCC,NF,E 621.

Formula: C₅H₈NNaO₄·H₂O
MW: 187.13 g/mol
Melting Pt: 225…240 °C
Flash Pt: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278

Order Now

Specification Test Results

Hill Formula C₅H₈NNaO₄ * H₂O
Molar Mass 187.13 g/mol
Flash point less than 900°C
Melting Point less than 200°C (sublimed)
pH value 6.7 - 7.2 (50 g/l, H₂O, 20 °C)
Bulk density 680 kg/m³
Solubility 600 g/l (20 °C)
LD 50 oral LD50 Rat 15800 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 1 slightly water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR