SilTite™ Metal Initial Installation Kits and Components, Gas Chromatography, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
073201 073202 073203 073220 073221 073222 073223 073226 073270 073271 073272 073300 073301 073302 073350 073351 073352 073400 073401 073450 073451 073452 073227 073204 073232 073205 073233 073231 073273
14226-370PK 242.1 CAD
14226-370 14226-372 14226-374 14226-376 14226-378 14226-380 14226-382 14226-388 14226-396 14226-398 14226-400 14226-402 14226-404 14226-406 14226-408 14226-410 14226-412 14226-420 14226-422 14226-424 14226-426 14226-428 14233-536 14233-668 14226-368 10799-736 10799-738 10799-740 10799-742 10799-744
SilTite™ Metal Initial Installation Kits and Components, Gas Chromatography, Trajan Scientific and Medical
Chromatography Supplies Chromatography Spare Parts
SilTite® metal ferrules have been developed specifically to overcome the problems associated with the use of 100% graphite and composite ferrules
  • Strong seal on capillary columns
  • Reduced air background
  • No contamination
  • Cleaner spectra
  • Leak free
  • Nut does not need re-tightening after initial temperature cycles.
SilTite ferrules form a strong and permanent airtight seal around the capillary column and have a temperature limit well above the temperature capacity of the injector, MS interface or GC oven. The metal SilTite ferrule together with the metalMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable