SilFlow™ Gas Chromatography Fittings, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
123720 123721 123722 123723 123724 123725 123730 123731 123732 123733 123734 123735 123704 123705 123706 123707 123709 123717 123742 123744 123755 123743 123750 123751 123752 123753
89205-338EA 1568.96 CAD
89205-338 89205-340 89205-342 89205-344 89205-346 89205-348 89205-350 89205-352 89205-354 89205-356 89205-358 89205-360 89205-362 89205-364 89205-366 89205-368 89205-370 89205-372 89205-374 89205-376 10799-718 10799-720 10799-722 10799-724 10799-726 10799-728 10799-730 10799-732
SilFlow™ Gas Chromatography Fittings, Trajan Scientific and Medical
Fittings and Connectors
SGE understands today’s chromatographers need to move from tubing based flow systems to planar microchannel systems to deliver flexible chromatography solutions
  • Chemically inert
  • Low dead volume
  • Superb operational stability
  • Easy to install
  • Leak free
These various fittings can be used for backflushing, column splitting, detector splitting, heart cutting, deans' switching and column switching. They are for use in multidimensional GC and anywhere a tee-piece has traditionally been used.
SilFlow™ is an innovation in design and fabrication resulting in a highly efficient and reliable micro fluidic platform that improves your GC connectivity to enable maximum chromatography performance.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Various configurations are available with ports to suit 0.25/0.32 mm and 0.53 mm I.D. columns, fused silica and 1.1 mm O.D. stainless steel tubing.