Water, Ultrapure for PCR, deionized, sterile

Supplier: QUALITY BIOLOGICAL, INC.

351-161-671 351-161-671EA 351-161-741
10128-566PK 712.83 CAD
10128-566 10128-568 10128-570
Water, Ultrapure for PCR, deionized, sterile
Water

PCR Water is useful for PCR Assays (i.e rRT-PCR, etc.).


  • DNase, RNase and Protease tested.
  • Free of contaminants of human, mouse and rat DNA.


PCR Qualified Water is functionally tested in a PCR reaction. PCR Qualified Water is highly purified water and it is reverse osmosis deionized, diethylpyrocarbonate (DEPC) treated. This product is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures. Free of detectable contamination of human, mouse and rat DNA.

Endotoxin and Mycoplasma Tested
Harmonization Code: 3822.00.5090
ECCN: EAR99


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR