Amphotericin B 250 µg/mL, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

120-097-711
10128-872EA 98.56 CAD
10128-872
Amphotericin B 250 µg/mL, sterile filtered
Amphotericin B

Amphotericin B prevents the growth of fungi by causing an increase in fungal plasma membrane permeability. Amphotericin B is used to prevent the contamination of cell cultures by yeast and multicellular fungi.


  • 100X concentration. Just dilute 1:100 to get the final working concentration of 2.50 µg/mL
  • Antifungal agent


Amphotericin B is useful for the maintenance of antifungal cell cultures in 2.5 mg/L concentration with penicillin and streptomycin also used in the medium.

Product Contents
Contains Amphotericin B; 250 µg/mL of deionized water

Recommendation
The recommended working concentration ranges from 0.25 to 2.50 µg/mL

UNSPSC: 41122100
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 24 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipped on dry ice.


Caution: Research Use Only

Formula: C₄₇H₇₃NO₁₇
MW: 924.08 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00877763
CAS Number: 1397-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR