Penicillin/Streptomycin/Amphotericin B mixture, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

120-096-711 120-096-711EA
10128-868PK 508.32 CAD
10128-868 10128-870
Penicillin/Streptomycin/Amphotericin B mixture, sterile filtered
Penicillin/Streptomycin/Amphotericin B

Amphotericin B, Penicilllin, Stretomycin Mix is useful for reducing the chance of microbial contamination. Recommended for use in cell culture applications at 10 mL/L.


  • Convenient mixture of the antibiotics and antifungal agent in one bottle.
  • 100X concentration, just dilute 1:100 in the culture medium


Penicillin-Streptomycin-Amphotericin B is a broad spectrum bacteriostatic and bactericidal, with activity against gram negative and gram-positive organisms including yeast and mold.Penicillin acts by inhibiting bacterial cell-wall synthesis. Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome and causes miscoding.

Amphotericin B interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out. Antimicrobial spectrum: Includes Gram-negative bacteria, Gram-positive bacteria, fungi and yeasts.

The antibiotic antimycotic solution of Amphoterecin B-Penicillin-Streptomycin is used to minimize the potential of microbial contamination when propagating primary cells or continuous cell lines in culture.

Application
Penicillin-Streptomycin-Amphotericin B Solution is used to reduce the chance of microbial contamination while propagating primary cells or continuous cell lines in culture. It is recommended for use in cell culture applications at 10 mL/L.

Product Contents
Contains Amphotericin B 25 µg/mL, Penicillin 10,000 Units/mL & Streptomycin 10,000 µg/mL

Cytotoxicity Tested
UNSPSC: 41122100
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 24 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipped on Dry Ice


Caution: Research Use Only. Storage Conditions : -10℃ to -20℃

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR