Penicillin : Streptomycin, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

120-095-671 120-095-721 120-095-721EA
10128-862PK 79.27 CAD
10128-862 10128-864 10128-866
Penicillin : Streptomycin, sterile filtered
Penicillin/Streptomycin

Penicillin-Streptomycin acts in two ways. Penicillin inhibits bacterial cell wall synthesis. While, streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome, causing miscoding.


  • 100X concentration of mixture of penicillin and streptomycin


Penicillin was originally purified from the fungi Penicillium and acts by interfering directly with the turnover of the bacterial cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to the 30S subunit of the bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria. This solution contains 10,000 units/mL of penicillin and 10,000 µg/mL of streptomycin.

UNSPSC: 41122100
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 24 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipped on dry ice


Caution: Research Use Only

Storage Temperature: Freezer
CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR