Cell Freezing Medium, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
It is important to store cells for future studies.
- Cell Freezing Medium A is a 2X concentration with Fetal Bovine Serum
- Cell Freezing Medium B is a 2X concentration with DMSO
Cell freezing medium A is frequently used in cell biology research for the cryopreservation of cells. Cell Freezing Medium A (2X) works with Cell Freezing Medium B (2X) at a 1:1 ratio.
Cryopreservation ensures that you have back-up cells in case of contamination and loss of cell supply. It is best to cryopreserve cells when they are at their maximum growth rate or almost confluent. Cell freezing medium A protects cells from damage after freezing and thawing.
Sterile filtered
Performance Tested
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: Accessories not included with Cell Freezing Medium
Caution: Research Use Only