SDS Protein Gel Loading Solution, 5X, Quality Biological
Supplier: Quality Biological
351-334-021 351-334-021EA
10128-592EA 143.81 CAD
10128-592 10128-590
SDS Protein Gel Loading Solution, 5X, Quality Biological
Buffers Electrophoresis Buffers Loading Buffers
Formulated as 5X rather than the traditional 2X stocks, SDS Protein Gel Loading Solution 5X enables a larger volume of sample to be loaded per well.
  • 5X concentrated protein gel loading buffer
Simply load the gel sample wells and visualize progress of the electrophoresis run by monitoring the location of the blue dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques, as well as Western blotting procedures.
SDS Protein Gel Loading Solution 5X is useful for SDS-PAGE and Western Blotting.
Contents: Tris base, 10% SDS, bromphenol blue, and glycerol
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.