SDS Protein Gel Loading Solution, 2X, Quality Biological

Supplier: QUALITY BIOLOGICAL, INC.

351-082-661 351-082-661EA
10128-538PK 596.18 CAD
10128-538 10128-540
SDS Protein Gel Loading Solution, 2X, Quality Biological
Buffers Electrophoresis Buffers Loading Buffers

"SDS Protein Gel Loading Solution, 2X is formulated at the traditional 2X stocks, for standard volume of sample to be loaded per well. Simply load the gel sample wells and visualize progress of the electrophoresis run by monitoring the location of the blue dye front.


  • Sterile filtered
  • Endotoxin and Performance tested


Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques.


SDS Protein Gel Loading Solution, 2X is a convenient and ready-to-use solution for SDS-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blotting.


Contents: Tris base, 4% SDS, bromphenol (bromophenol) blue 0.2%, glycerol 20%, and molecular biology grade water.


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Ordering information: Optional - add dithiothreitol (0.2ml 1M DTT per ml) or 2-mercaptoethanol (0.02 ml 2-ME per ml) immediately before use.


Caution: Research use only.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR