RNA Gel Loading Solution 10X, Quality Biological

Supplier: QUALITY BIOLOGICAL, INC.

351-081-661 351-081-661EA
10128-534PK 528.45 CAD
10128-534 10128-536
RNA Gel Loading Solution 10X, Quality Biological
Buffers Electrophoresis Buffers Loading Buffers

RNA Gel Loading Solution, 10X is prepared according to Sambrook, Fritsch & Maniatis (1989)


  • Contains two dyes, bromphenol blue & xylene cyanole FF
  • Filtered through a 0.22 µm membrane
  • RNase tested


The purpose of this reagent is to ensure that the RNA samples remain submerged in the wells of the gel, by making them denser than the electrophoresis buffer.


RNA Gel Loading Solution is suitable for RNA Gel Electrophoresis and Northern Blot Analysis.


Contents: Bromphenol blue 0.25%, xylene cyanole FF 0.25%, disodium EDTA dehydrate, glycerol 50%, and DEPC treated water


DNase, RNase and Protease Tested.


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Caution: Research use only.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR