Trypsin Delarco
Supplier: QUALITY BIOLOGICAL, INC.
118-150-721 118-150-721EA
10128-798PK 246.37 CAD
10128-798 10128-800
Trypsin Delarco
Enzymes
Trypsin Delarco assists in the process of cell culture and transfection.
  • With HBSS without calcium and magnesium
  • Without Phenol Red
Trypsin is a proteolytic enzyme used to detach adherent cells from culture vessel surfaces. Trypsin delarco is used in cell passaging, to ensure stable products.
Sterile Filtered
Concentration: 0.1 mM disodium EDTA
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months
Certifications: Manufactured in an ISO 9001:2015 certified facility.
Ordering information: *NOTE: During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not effect the functionality of the trypsin.
Caution: Research Use Only