Trypsin, 2.5%
Supplier: QUALITY BIOLOGICAL, INC.
118-086-721 118-086-721EA
10128-780PK 353.57 CAD
10128-780 10128-782
Trypsin, 2.5%
Enzymes
Trypsin is a proteolytic enzyme used to detach adherent cell from culture vessel surfaces. Typical use includes removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.
  • Without EDTA
  • Without Phenol Red
Trypsin is a serine protease derived from porcinepancreas. It is a single chain polypeptide of 223 amino acid residue with substrate specificity based on positively charged Lysine and Arginine side chains. Trypsin predominantly cleaves peptide chains at the carboxyl sides of Lysine and Arginine, except when either is followed by Proline. It is most commonly used for dissociation and disaggregation of adherent cells.
Recommendation
Do not add calcium or magnesium.
Prolonged exposure can negatively affect cell viability. Be aware of exposure time.
Sterile Filtered
Performance Tested
Certifications: Manufactured in an ISO 9001:2015 certified facility.
Ordering information: *NOTE: During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not effect the functionality of the trypsin.
Caution: Research Use Only