Sodium bicarbonate solution 7,5% (0.893 mol/L)

Supplier: QUALITY BIOLOGICAL, INC.

Synonyms: Sodium bicarbonate

118-085-721 118-085-721EA
10128-776PK 306.19 CAD
10128-776 10128-778
Sodium bicarbonate solution 7,5% (0.893 mol/L)
Sodium hydrogen carbonate

Sodium Bicarbonate (NaHCO₃) is typically used in cell culture media as a buffer.


  • Endotoxin and Mycoplasma Tested
  • Osmolality: 290 ± 5% mOsM/kg H₂O
  • Sterile Filtered (0.1 μm)


Although NaHCO3 does not have the buffering capacity of HEPES, it is occasionally preferred over HEPES because of both its lower toxicity and nutritional value to cells in culture. Directions: Dilute the Sodium Bicarbonate 7.5% (0.893M) in the appropriate cell culture medium. Typically, Sodium Bicarbonate is diluted to a final concentration of 5-15 mM.

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR