Sodium pyruvate 100 mM in water (100 X) cell culture reagent, sterile-filtered

Supplier: QUALITY BIOLOGICAL, INC.

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

116-079-721 116-079-721EA
10128-764PK 225.42 CAD
10128-764 10128-766
Sodium pyruvate 100 mM in water (100 X) cell culture reagent, sterile-filtered
Sodium pyruvate

Sodium pyruvate is used by cells as an easily accessible carbohydrate source. Additionally, it is involved with amino acid metabolism and initiates the Kreb′s cycle. The 100 mM solution should be diluted 1:100 for most cell culture applications.


  • 100 mM solution
  • Sterile Filtered


Sodium pyruvate solution has been used in different type of cell culture media.


Recommendation
Protect from light. Add to cell culture media as a carbon source at 1 mM or 100 mg/mL.
Max Shelf Life: 24 months from date of manufacture


Harmonization Code: 3822.00.5090
ECCN: EAR99


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Caution: Research Use Only

Formula: CH₃COCOONa
Storage Temperature: Refrigerator
MDL Number: MFCD00002586
CAS Number: 113-24-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR