SOC Medium, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
340-031-671 340-031-671EA
10128-288PK 362.17 CAD
10128-288 10128-290
SOC Medium, Quality Biological
Cell Culture Media
SOC medium is a ready-to-use solution, useful in the final step of bacterial cell transformation to obtain maximal transformation efficiency of E. coli.
  • Without magnesium chloride, needs to be added before use.
SOC is a microbial growth medium used for the transformation of competent cells (E. coli). This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and glucose in a low-salt formulation.
More specifically SOC Medium can be used: in the propagation of DH-alpha (of E. coli strains) primarily in the recovery step of Escherichia coli competent cell transformations, and to maximize the transformation efficiency of competent cells.
Recommendation
Immediately before use, add 2M magnesium chloride to a final concentration of 20mM (10ml MgCl2/L).
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Order Now
SPECIFICATIONS
Environmentally Preferable
Sterility Autoclave sterilized
Shipping conditions Ambient temperature