LB Agar 1.5% (Lennox), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
340-025-211 340-025-231
10128-280CS 1160.75 CAD
10128-280 10128-282 10128-284 10128-286
LB Agar 1.5% (Lennox), Quality Biological
Media for Microbiology Prepared Media for Microbiology
LB Agar 1.5% Lennox provides all nutritional requirements of these organisms. LB Agar 1.5% Lennox contains half the sodium chloride level of the Miller formulation of LB Agar, allowing the researcher to select the optimal salt concentration for a specific strain.
  • Just autoclave to melt and pour.
LB Agar 1.5% Lennox is nutritionally rich. It was developed by Lennox for growth and maintenance of pure culture of recombinant strains of E. coli. These strains are generally derived from E. coli K12, which are deficient in B vitamin production. This strain of E. coli has been further modified through specific mutation to create an auxotrophic strain that is not capable of growth on nutritionally deficient media.
LB Agar 1.5% is useful for propagation of bacteriological stocks, as well as plating and screening bacteriological clones.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only