Water nuclease-, protease-free, low endotoxin, deionized, sterile, DEPC treated

Supplier: QUALITY BIOLOGICAL, INC.

351-068-131 351-068-131CS 351-068-151 351-068-161 351-068-491 351-068-721 351-068-721EA
10128-514EA 146.02 CAD
10128-514 10128-516 10128-518 10128-520 10128-522 10128-524 10128-526
Water nuclease-, protease-free, low endotoxin, deionized, sterile, DEPC treated
Water

DEPC Treated Water is Ultra Pure and prepared by reverse osmosis, passed through fine carbon, de-ionized through two resin beds and serially filtered 1 µm positively charged membranes. The water is subsequently treated with 0.1% (v/v) DEPC (Diethylpyrocarbonate) and incubated and heated overnight at 37°C. Finally, DEPC Treated Water is filtered through a 0.1 µm filter.


  • DNase, RNase and Protease tested


DEPC Treated Water can be used to reconstitute nucleic acids and/or proteins. In addition, it can be used as a diluent or solvent for any molecular biology grade reagent. DEPC Treated Water may be usefully employed in RNA Applications (e.g., Preparations of an RNA probe, DNA Applications (e.g., Subcloning in pUC plasmids), and Protein Applications (e.g., Western Blotting).

Harmonization Code: 3822.00.5090
ECCN: EAR9


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR