LB Broth (Lennox), Quality Biological

Supplier: QUALITY BIOLOGICAL, INC.

340-016-101 340-016-101CS
10128-256EA 83.31 CAD
10128-256 10128-258 10128-260 10128-262 10128-264 10128-266
LB Broth (Lennox), Quality Biological
Media for Microbiology Prepared Media for Microbiology

LB Broth, Lennox is a ready-to-use microbial growth medium used for the cultivation of E. coli.


  • Low (5 g/L) salt formulation
  • Performance Tested


LB Broth is a low-salt formulation in comparison to its counterpart LB Broth, Bertani Miller. Lennox is also nutrient rich containing peptides, amino acids, water soluble vitamins and carbohydrates.LB Broth, Lennox is useful for molecular genetic studies, as well as maintenance and propagation of bacteriological stocks.


Recomendation
This is the best option for LB Broth if a lower concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl.


UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99


Certifications: Manufactured in an ISO 9001:2015 certifed company in accordance with ISO-14644 and IEST-RP-006.3; meeting standards for ISO Class 8 and ISO Class 7.


Caution: Research Use Only

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR