SSC, 20X Buffer, Quality Biological

Supplier: QUALITY BIOLOGICAL, INC.

351-003-101 351-003-131
10128-688EA 80.45 CAD
10128-688 10128-690 10128-692 10128-694
SSC, 20X Buffer, Quality Biological
Buffers Electrophoresis Buffers Hybridisation Buffers

SSC 20X buffer is a ready-to-use standard reagent in Southern and Northern hybridization procedures used to control stringency for the washing steps after hybridization


  • Supplied in 1 L plastic bottles, or in a 4 L or 10 L stackable Cubitainer® Box
  • Ready-to-use
  • Economical
  • Filtered through a 0.22 µm membrane
  • DNase, RNase, and Protease tested


SSC 20X is used for several Cell Biology applications such as: preparing DNA and RNA hybridization buffers and post-hybridization membrane washing buffers, hybridization buffer for Northern and Southern blots, and as a washing buffer for Northern and Southern blots.


The concentrated formula can be used straight or diluted to make SSC working solutions with concentrations as low as 0.5X.


Contents: Sodium chloride, sodium citrate dihydrate, molecular biology grade water


Certifications: Manufactured in an ISO 9001:2015 certifed company in accordance with ISO-14644 and IEST-RP-006.3; meeting standards for ISO Class 8 and ISO Class 7.


Ordering information: Please call a Avantor sales representative if you plan to order more than 100 bottles


Caution: Research use only.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR