Sodium acetate 3 M in aqueous solution, sterile filtered pH 5.2
Supplier: QUALITY BIOLOGICAL, INC.
Synonyms:
Acetic acid sodium salt
Sodium Acetate, 3M pH 5.2 is a common molecular biology reagent. All production lots are tested to ensure that they are free of Dnase, Rnase & Proteases. It is often used in buffer systems to maintain the pH of a given solution.
- Filtered through a 0.22 µm membrane
- Dnase, Rnase, and Protease tested
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Delivery information: Shipping Condition: ambient
Caution: Research Use Only
Formula:
H₃CCOONa Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...