Gentamicin sulfate 50 mg/mL, sterile filtered

Supplier: QUALITY BIOLOGICAL, INC.

Synonyms: Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

120-098-661EA 120-098-661
10128-216EA 147.86 CAD
10128-216 10128-874
Gentamicin sulfate 50 mg/mL, sterile filtered
Gentamicin sulphate

Gentamicin is an aminoglycoside complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is used as the sulfate salt.


  • Ready to use solution


Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria. Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination on cell cultures. Antimicrobial spectrum: Includes Gram-negative and Gram-positive bacteria, including strains resistant to tetracycline, chloramphenicol, kanamycin and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus and Streptococcus.

Gentamicin sulfate is a broad-spectrum antibiotic used as a selection agent (gentamicin-resistance gene) in molecular biology and cell culture applications. This product is formulated to contain 50 mg/mL Gentamicin in deionized water and is recommended for use in eukaryotic cell culture at a volume of 1 milliliter per liter.

UNSPSC: 41122100
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS Number: 1405-41-0
Merck Index: 13,04403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR