Water for molecular biology nuclease-, protease-free, low endotoxin, deionised, sterile
Supplier: QUALITY BIOLOGICAL, INC.
Synonyms:
Hydrogen oxide
Molecular Biology Water is prepared by reverse osmosis, passed through fine carbon, deionized through two resin beds, and serially filtered through 0.1 µm positively charged membranes.
- DNase, RNase and protease tested
Molecular Biology Water is ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase, DNase, and Protease-free water is required.
No toxic agents, including DEPC, are used in the preparation of Molecular Biology Water, eliminating possible interferences with enzymatic reactions. Molecular Biology Water is negative for RNase (sensitivity to femtomoles) and is negative for non-specific Endonuclease, Exonuclease and Protease activity.
Molecular Biology Water is useful in: reconstituting nucleic acids and/or proteins, RNA applications (i.e. Preparation of an RNA probe), DNA applications (i.e. Subcloning in pUC plasmids), and protein applications (i.e. Western Blotting). Molecular Biology Water can also be used as a diluent or solvent for any molecular biology grade biochemical.
UNSPSC: 41116134
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Delivery information: Shipping Condition: ambient
Caution: Research Use Only
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Density: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...