DNA Gel Loading Solution, 5X, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
351-028-661 351-028-661EA
10128-448PK 394.13 CAD
10128-448 10128-450
DNA Gel Loading Solution, 5X, Quality Biological
Buffers Electrophoresis Buffers Loading Buffers
DNA Gel Loading Solutio, 5X is prepared according to Sambrook, Fritsch & Maniatis. The purpose of this reagent is to be a tracking dye during electrophoresis. It ensures that DNA samples remain submerged in the wells of the gel, by making them denser than the electrophoresis buffer.
  • Ready-to-use format
  • Two-color tracking of DNA migration during DNA electrophoresis
  • 5X concentration
  • Filtered through a 0.22 µm membrane
  • Dnase, Rnase, and Protease tested
The dye in DNA Gel Loading Solution has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition.
DNA Gel Loading Solution, 5X is suitable for: use with agarose or non-denaturing polyacrylamide gel electrophoresis (PAGE), DNA gel electrophoresis, Northern and Southern blot analysis, and most commonly loading and tracking of DNA samples on electrophoretic gels.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.