Tryptic Soy Agar Plates with 5% Sheep Blood, Quality Biological
Supplier: Quality Biological
441-120-231
10128-598CS 1459.05 CAD
10128-598
Tryptic Soy Agar Plates with 5% Sheep Blood, Quality Biological
Media for Microbiology Prepared Media for Microbiology
Trypticase Soy Agar with 5% Sheep Blood is used for cultivating fastidious microorganisms and for the visualization of hemolytic reactions produced by many bacterial species
Grow a wide variety of fastidious organisms on TSA with Sheep Blood, a general purpose non-selective medium with the addition of sheep blood in the formulation.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only