Ampicillin 100 mg/mL ready-to-use, sterile filtered
Supplier: QUALITY BIOLOGICAL, INC.
351-344-731
10128-594PK 351.58 CAD
10128-594
Ampicillin 100 mg/mL ready-to-use, sterile filtered
Ampicillin
Ampicillin in a convenient ready-to-use solution useful in selecting for ampicillin resistant bacteria. Ampicillin is a β-lactam antibiotic with an amino group side chain attached to the penicillin structure.
  • Convenient 1000X concentrated stock
  • Small 1 mL size to prevent freeze-thaw cycle
  • Affordable price
Ampicillin, which is a semi-synthetic derivative of penicillin, inhibits cell wall synthesis (peptidoglycan crosslinking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane in Gram-negative and Gram-positive bacteria.2,3 Cleavage of the β-lactam ring of ampicillin by β-lactamase results in bacterial resistance to this antibiotic.
Product Contents
Ampicillin and Molecular Biology Grade Water
Recommendation
The recommended concentration for antibacterial use in culture media is ~100 mg/mL (a 1000-fold dilution of the product). The recommended concentration for use in ampicillin resistance studies is 20–125 mg/mL.
Functional Tested
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 24 months
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Delivery information: Shipping Condition: on dry ice
Caution: Research Use Only