HyFlex® 11-927 Oil-Repellent Gloves with Polyethylene Liner, 3/4 Dipped, Ansell
Supplier: Ansell Canada
111734 111735 111736 111737 111738 111739
CA10049-056CS 3351.97 CAD
CA10049-056 CA10049-058 CA10049-060 CA10049-062 CA10049-064 CA10049-066
HyFlex® 11-927 Oil-Repellent Gloves with Polyethylene Liner, 3/4 Dipped, Ansell
Gloves
Combining advanced cut resistance, oil repellence, and oil grip in a single glove, the Hyflex 11-927® features Ansell’s RIPEL™ liquid-repellent technology with durable double coating, along with the industry-leading Ansell Grip™ technology on the palm, for a secure grip in oily environments.
  • Effective oil-wicking microscopic channels
  • Strong polyethylene fiber provides excellent cut protection
  • Best-in-class comfort
  • No double gloving necessary
  • Reduced back-of-hand abrasions and snags
This patented grip feature uses microscopic channels to wick away oil and moisture from the surface of parts or tools, ensuring safe, secure, and effective glove contact.
Glove liner is made of a light, strong high-performance polyethylene (HPPE) fiber that provides ANSI Level 2 (EN 3) cut protection, with a 3/4 dip primary coating. Glove design combines the abrasion resistance of two coatings: the 3/4 nitrile dip primary layer and the nitrile grip palm dip coating. The unique combination of polymers provides oil repellence in a 3/4 dip geometry, eliminating double gloving and reducing back-of-hand abrasion and snags. This unique blend of protection and performance makes the 11-927 a versatile choice for any application requiring handling of sharp metal components coated in oil or other lubricants.
Diverse HyFlex styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.
RIPEL™ technology creates a liquid-impermeable layer in knit glove styles that prevents oil or lubricant from making even incidental contact with the wearer’s skin, while also enhancing hand health. Ansell Grip™ technology is a coating treatment that minimizes the force required to grip oily or wet tools or materials, reducing hand and arm fatigue and improving dexterity, safety and productivity.
Certifications: Abrasion (ANSI Level 4) and cut (ANSI Level 2) resistant.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.