Brucella Broth (BLBB) with GSS Supplement Plates, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
340-317-211 340-317-231
10128-378CS 1554.81 CAD
10128-378 10128-380
Brucella Broth (BLBB) with GSS Supplement Plates, Quality Biological
Media for Microbiology Prepared Media for Microbiology
Bisulfateless Brucella Plate (BLBB) is applicable for use in the primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms from clinical specimens.
  • Special formulation without bisulfate
BLBB with GSS Supplements is a non-selective medium suitable for the growth of aerobic and microaerophilic bacteria when incubated under the appropriate conditions. BLBB Plates are useful for the enrichment, isolation and cultivation of Brucella species.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: NA
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only