1/2 YPD Plates, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
YPD is a media variant derived from YPAD. YPAD consists of nutrient base (yeast extract and peptone), adenine supplement and a carbon source. YPD’s formulation is the same but without the additional adenine.
YPD 1/2 Plates are useful for subculturing of untransformed yeast, yeast two-hybrid screening, yeast one-hybrid screening, enrichment and isolation of yeast; ade2 yeast will turn red on YPD.
- Ready to use in petri dishes
- Packed in 10 plates per sleeve
- Re-closable zipper pouch for convenient storage
For routine subculturing of untransformed yeast
Yeast two-hybrid screening
Yeast one-hybrid screening
Enrichment and isolation of yeast
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: NA
Certifications: Manufactured in an ISO 9001:2015 certifed company in accordance with ISO-14644 and IEST-RP-006.3; meeting standards for ISO Class 8 and ISO Class 7.
Caution: Research Use Only