SilTite™ FingerTite GC Ferrule Systems, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
1034085 073610 073612 073620 073621 073628 073629 073630 073631 073633 073636 073640 0736100 0736101 0736102 0736103 0736104 0736105 07361053 07361054 073611 073614 073615 073616 073618 073619 07362053 073622 073623 073625 073632 073554 073473 073230 073330 073331 073332 073219 073228 073229
14226-536PK 120.65 CAD
14226-536 97051-946 97051-948 97051-950 97051-952 97051-954 97051-956 97051-958 97051-960 97051-962 97051-964 97051-966 89232-136 89232-140 89232-142 89232-144 89232-146 89232-148 89232-150 89232-152 89232-154 89232-160 89232-166 89232-168 89232-170 89232-176 89232-178 89232-182 89232-188 89232-190 89232-196 10799-794 10799-796 10799-810 10799-812 10799-814 10799-816 10799-820 10799-822 10799-824
SilTite™ FingerTite GC Ferrule Systems, Trajan Scientific and Medical
Fittings and Connectors
SilTite FingerTite is a GC ferrule system that has been designed to be installed only with the force of you fingers, delivering an easy, leak and maintenance free seal.
  • Simplified installation process
  • Leak free
  • No column creep
  • Cannot overtighten
  • No broken columns
  • Chemically inert
  • No contamination
  • Metal ferrules
  • No temperature limit
  • No tools required
These ferrules are for use with MS, high throughput laboratories. Starter kits are available for Agilent, PerkinElmer, Shimadzu, Thermo Scientific and Varian/Bruker MC-MS systems.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Each starter kit includes all the parts necessary to convert one GC system (one injector and one detector) to the SilTite FingerTite system. In addition there are 5 SilTite FingerTite nuts, one packet (10 ferrules) of 0.4 mm ID SilTite FingerTite ferrules (suitable for installing 0.1 - 0.25 mm ID capillary columns) and a ferrule install tool which allows you to seat the ferrule in the correct position on the capillary column.