Qualitative Filter Paper, Grade 613, Ahlstrom
Supplier: Ahlstrom
6130-0606 6130-0808 6130-0470 6130-1250 6130-1100 6130-1500 6130-0425 6130-0550 6130-0750 6130-0900 6130-1850
28340-010PK 370.23 CAD
28340-010 28342-022 28342-054 71440-012 71440-010 71440-014 71440-026 71440-028 71440-030 71440-032 71440-017
Qualitative Filter Paper, Grade 613, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom Grade 613 is a strong general-purpose filter with a medium flow rate and retention.
It is commonly used in student laboratories. This low ash grade contains more than 95% super refined alpha-cellulose and linter fibers.