Glass Microfiber Filter Paper, Grade 169, Ahlstrom
Supplier: Ahlstrom
1690-0210 1690-0240 1690-0425 1690-0470 1690-0550 1690-0700 1690-0900 1690-1100 1690-1250
10199-604PK 114.11 CAD
10199-604 10199-606 10199-608 10199-610 10199-612 10199-614 10199-616 10199-626 10199-628
Glass Microfiber Filter Paper, Grade 169, Ahlstrom
Filters Glass Fiber Filters
Grade 169 is a microfiber glass grade formulated for product stability when heated over 500 degrees Centrigrade, as required for testing for volatile solids in STM 2540.