Quartz Extraction Thimbles, Ahlstrom
Supplier: Ahlstrom
E360-4323
10199-580PK 1499.42 CAD
10199-580
Quartz Extraction Thimbles, Ahlstrom
Extractors Extraction Thimbles
Quartz thimbles are used for emission testing in high temperature environments
Wall thickness is approximately 2 mm thick. Temperature resistance of 900 degrees Celsius.