ReliaDisc™ Sterile Cellulose Nitrate Gridded Membrane, Ahlstrom
Supplier: Ahlstrom
760224 760219
10192-154PK 1571.86 CAD
10192-154 10192-174 10192-176
ReliaDisc™ Sterile Cellulose Nitrate Gridded Membrane, Ahlstrom
Filters Membrane Filters
Ahlstrom offers gridded membranes for use in colony counting, particule testing, and microscopy.
These cellulose nitrate membranes are gridded and available either packaged in individual sleeves or in a dispenser strip. The dispenser strip is usable in standard equipment from various manufacturers.