pH Indicator Papers and Strips, Ahlstrom
Supplier: Ahlstrom
PHT1-0014 PHR5-0114 PHB2-0114 PHR5-0111 PHR3-0111
10199-582PK 34.32 CAD
10199-582 10199-598 10199-600 10199-618 10199-620 10199-622
pH Indicator Papers and Strips, Ahlstrom
Test Papers pH Indicator Papers and Test Strips
Ahlstrom offers a range of test papers for rapid determination of pH
Universal pH Indicator papers are availabe in a roll of 5 m in a plastic case, a refill pack of 3 reels for that case, and a box of 200 strips. pH test strips, which have small squares of paper, each prepared with a different indicator for more accurate determination of pH are also available. All include color charts.