Syringes, Diamond MS, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
002750 002760 002770 002780 002790 003750 003760 003770 003790 004750 004770 004790 005750 005770 005790 006750 007750 0355210 0355211 0355213 0355215 0355216 0355220 0355221 0355223 0355225 0355226 0355230 0355231 0355233 0355235 0355236 0355310 0355311 0355312 0355313 0355320 0355321 0355322 0355323 0355330 0355331 0355332 0355333 0355410 0355411 0355420 0355421 0355430 0355431 0355510 0355511 0355512 0355520 0355521 0355522 0355530 0355531 0355532 0355610 0355611 0355612 0355620 0355621 0355622 0355630 0355631 0355632
89236-156EA 229.15 CAD
89236-156 89236-158 89236-160 89236-162 89236-164 89236-200 89236-202 89236-204 89236-206 89236-216 89236-218 89236-220 89236-230 89236-232 89236-234 89236-248 89236-262 89236-364 89236-366 89236-368 89236-370 89236-372 89236-374 89236-376 89236-378 89236-380 89236-382 89236-384 89236-386 89236-388 89236-390 89236-392 89236-394 89236-396 89236-398 89236-400 89236-402 89236-404 89236-406 89236-408 89236-410 89236-412 89236-414 89236-416 89236-418 89236-420 89236-422 89236-424 89236-426 89236-428 89236-430 89236-432 89236-434 89236-436 89236-438 89236-440 89236-442 89236-444 89236-446 89236-448 89236-450 89236-452 89236-454 89236-456 89236-458 89236-460 89236-462 89236-464
Syringes, Diamond MS, Trajan Scientific and Medical
Syringes Syringes for Chromatography
SGE understands how important your MS results are and how precious your sample is, so we engineered new syringes and needles to bring you the Diamond MS Syringe range. Diamond MS syringes and needles are MS ready and designed to improve the sensitivity of your MS results.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment
The choice for sensitivity–how to choose your MS ready syringe and needle for improved sensitivity: 1. Choose a manual syringe or one specifically designed for your instrument. 2. Select the syringe capacity. 3. Choose the needle geometry – LC or GC injections, tip design and needle dimensions. 4. Choose which 'Ultra Smooth' needle best suits your application.
Accuracy and Reproducibility are ± 1 % (displaced volume). Temperature Range is 5-120 °C. International Standards Traceability.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Available in 10 to 500 µL manual syringes 10 to 100 µL CTC and Thermo RSH autosampler syringes 10 and 25 µL Agilent autosampler syringes 10 µL Shimadzu autosamplers syringes. Ultra Smooth Ultra Smooth Hydrophilic and Ultra Smooth Hydrophobic needles.