Protector® XL™ Floor-Mounted Laboratory Hoods with Horizontal Sashes, Labconco
Supplier: Labconco
112816002 112826002 112836002 112016002 112026002 112036002 112116002 112126002 112136002 112216002 112226002 112236002
10221-900EA 41001 CAD
10221-900 10221-902 10221-904 10221-906 10222-154 10222-156 10222-158 10222-160 10222-162 10222-164 10222-166 10222-168
Protector® XL™ Floor-Mounted Laboratory Hoods with Horizontal Sashes, Labconco
Fume Hoods and Enclosures Chemical Fume Hoods
Protector XL floor-mounted laboratory hoods with horizontal-sliding sashes are suitable for general chemistry applications and have the dimensions required for over-sized equipment.
These hoods are constructed of durable, powder-coated steel with four tempered safety glass sashes and a corrosion- and chemical-resistant, fiberglass-reinforced composite panel liner that surpasses all national codes for flame spread. Features include removable access panels, 73.5" loading height and pre-wired LED lighting rated for 50000 hours.
Laboratory workers with applications requiring distillation racks, extraction apparatus or large drums and carboys benefit from the increased loading height these hoods provide.
Certifications: Protector XL floor-mounted laboratory hoods with horizontal-sliding sashes are performance tested to ASHRAE 110-1995, ETL-listed and CE approved. See our brochure for a complete list of regulations and standards to which these models conform.
Ordering information: All models require a remote blower and ductwork, not included. Models are available in widths of 8', 10', 12' and 16' and depths of 37.7", 43.7" and 55.7". These hoods can be ordered with 0 or 2 service fixtures, or 2 service fixtures and 2 GFCI Duplex receptacles. Electrical options of 100 to 115 volts, 50/60 Hz, 10 A or 208 to 230 volts, 50/60 Hz, 5 amps are offered. Hoods are shipped in multiple sections: top, bottom, and sashes. Protector XL floor-mounted hoods with vertical-rising sashes are also available. Contact your VWR representative for more information.