VWR® Cell Strainers, DNase/RNase Free, Non-Pyrogenic, Sterile
Supplier: VWR International
10199-654 10199-656 10199-658 10199-655 10199-657 76327-098 76327-100 76327-102
10199-654CS 209.54 CAD
10199-654 10199-656 10199-658 10199-655 10199-657 76327-098 76327-100 76327-102
VWR® Cell Strainers, DNase/RNase Free, Non-Pyrogenic, Sterile
Cell Strainers
Cell strainers are manufactured from a strong nylon mesh with evenly spaced mesh pores.
  • Ready-to-use, sterilized by electron beam irradiation
  • Individually packaged
  • Fit into most 50 mL conical tube
  • DNase- and RNase-free
  • Non-pyrogenic
These cell strainers are sterile, easy-to-use devices for quickly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues
Available in 3 mesh sizes; 40µm, 70µm, and 100µm. Also available in 3 different colors; blue, white, and yellow, for easy identification. Improved uniformity ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M