MultiLab IDS Conductivity and Temperature Sensors, Four-Granite Electrodes, YSI
Supplier: YSI
301710Y 301711Y
10066-688EA 1012.75 CAD
10066-688 10066-690
MultiLab IDS Conductivity and Temperature Sensors, Four-Granite Electrodes, YSI
Electrodes Conductivity Sensors
The IDS sensors automatically store their unique serial number and calibration data.
In addition, they also digitally process the measurement signal. The IDS sensors can be moved from instrument to instrument and maintain their calibration data and transmit that information to the new instrument.
Ordering information: Sensors come with a two year warranty.