YSI MultiLab IDS pH and Temperature Sensors, YSI
Supplier: YSI
103741Y 103750Y 103780Y 103740Y
10066-680EA 572.86 CAD
10066-680 10066-682 10066-684 10067-044
YSI MultiLab IDS pH and Temperature Sensors, YSI
Electrodes pH Electrodes
The IDS sensors automatically store their unique serial number and calibration data.
In addition, they also digitally process the measurement signal. The IDS sensors can be moved from instrument to instrument and maintain their calibration data and transmit that information to the new instrument.
Ordering information: Sensors come with a one year warranty.