HyFlex® 11-600B Black Nylon Gloves with Gray Coated Palm, Ansell
Supplier: Ansell Healthcare
205650 205651 205652 205653 205654
80092-472CS 1137.38 CAD
80092-472 80092-474 80092-476 80092-478 80092-480
HyFlex® 11-600B Black Nylon Gloves with Gray Coated Palm, Ansell
Gloves
Delivering bare-handed dexterity with the protection of a glove, these products provide a relaxed, ergonomic fit.
  • Durable construction
  • Machine washable
  • Maximum dexterity and sensitivity
  • Relaxed, ergonomic fit
Gloves have performance characteristics suited to a wide variety of applications and work environments.
The glove’s solid gray polyurethane coating hides soil, providing longer wear and reducing overall replacement costs. The durable gloves can be machine washed and reused with minimal shrinking and excellent recovery. Maximum dexterity and sensitivity for jobs that require handling or assembling small parts.
Diverse HyFlex® styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.
Use in the automotive and transportation, machinery and equipment, and maintenance industries. Uses include assembly of small pieces such as small bearings, joints and springs.
Certifications: ANSI Abrasion Level 3.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.